Product Details
- SNP ID
-
rs140261687
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:146639236 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCAAGTACTCGGCACTGCACAGCAC[C/T]TCGCCGGGCTCCTCGGCTCCCATCG
- Phenotype
-
MIM: 113725
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
POU4F2
PubMed Links
Gene Details
- Gene
- POU4F2
- Gene Name
- POU class 4 homeobox 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_004575.2 |
344 |
Silent Mutation |
ACC,ACT |
T32T |
NP_004566.2 |
View Full Product Details