Product Details

SNP ID
rs140716884
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:166734429 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTTTGTTTTTCTTGCCATGTGGT[A/G]TCTCTCTGTTATCCTGACTCAACTA
Phenotype
MIM: 607989
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SPOCK3 PubMed Links

Gene Details

Gene
SPOCK3
Gene Name
sparc/osteonectin, cwcv and kazal-like domains proteoglycan (testican) 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040159.1 1830 UTR 3 NP_001035249.1
NM_001204352.1 1830 UTR 3 NP_001191281.1
NM_001204353.1 1830 UTR 3 NP_001191282.1
NM_001204354.1 1830 UTR 3 NP_001191283.1
NM_001204355.1 1830 UTR 3 NP_001191284.1
NM_001204356.1 1830 UTR 3 NP_001191285.1
NM_001204358.1 1830 Intron NP_001191287.1
NM_001251967.1 1830 UTR 3 NP_001238896.1
NM_016950.2 1830 UTR 3 NP_058646.2
XM_011532018.1 1830 UTR 3 XP_011530320.1
XM_017008257.1 1830 UTR 3 XP_016863746.1
XM_017008258.1 1830 UTR 3 XP_016863747.1

View Full Product Details