Product Details

SNP ID
rs141597421
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:154604877 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAAAGTGAGTCATATTCTGTTTCC[A/G]CAGGGTGCTCTGGTCTGACCTGTTT
Phenotype
MIM: 134850
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FGG PubMed Links

Gene Details

Gene
FGG
Gene Name
fibrinogen gamma chain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000509.5 1460 Intron NP_000500.2
NM_021870.2 1460 Missense Mutation NP_068656.2

View Full Product Details