Product Details
- SNP ID
-
rs142420378
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:39406967 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGTGGCAAATGAAGCCAGGCTGTGC[G/T]GCAGGATCTCCAGGGAATGAATGGA
- Phenotype
-
MIM: 611135
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
KLB
PubMed Links
Gene Details
- Gene
- KLB
- Gene Name
- klotho beta
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_175737.3 |
115 |
Silent Mutation |
GCG,GCT |
A6A |
NP_783864.1 |
- Gene
- MIR5591
- Gene Name
- microRNA 5591
There are no transcripts associated with this gene.
View Full Product Details