Product Details

SNP ID
rs142878827
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:88710154 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCAATCCTTATGAGACCAGACTTG[C/G]AAAAATTACAGTAACAGAGTGAAAA
Phenotype
MIM: 613300 MIM: 605200
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FAM13A-AS1 PubMed Links

Gene Details

Gene
FAM13A-AS1
Gene Name
FAM13A antisense RNA 1
There are no transcripts associated with this gene.

Gene
HERC3
Gene Name
HECT and RLD domain containing E3 ubiquitin protein ligase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271602.1 Intron NP_001258531.1
NM_001318505.1 Intron NP_001305434.1
NM_014606.2 Intron NP_055421.1
XM_005263327.3 Intron XP_005263384.1
XM_005263329.3 Intron XP_005263386.1
XM_005263330.3 Intron XP_005263387.1
XM_005263331.1 Intron XP_005263388.1
XM_005263332.3 Intron XP_005263389.1
XM_011532397.2 Intron XP_011530699.1
XM_017008806.1 Intron XP_016864295.1
XM_017008807.1 Intron XP_016864296.1
XM_017008808.1 Intron XP_016864297.1

View Full Product Details