Product Details

SNP ID
rs143160739
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:44174993 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCTGAAAGAGTTCACTGTTTCTGC[A/G]TTTGTCTGGTGGGGGTATCCATTGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KCTD8 PubMed Links

Gene Details

Gene
KCTD8
Gene Name
potassium channel tetramerization domain containing 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198353.2 1503 Missense Mutation CGC,TGC R407C NP_938167.1
XM_011513690.2 1503 Missense Mutation CGC,TGC R435C XP_011511992.1

View Full Product Details