Product Details
- SNP ID
-
rs143183155
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:182891404 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAGATTGAATGAGATGTAACTCACT[C/G]AAGCTTTTGACTCGGTCTGCTGTTA
- Phenotype
-
MIM: 607638
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
DCTD
PubMed Links
Gene Details
- Gene
- DCTD
- Gene Name
- dCMP deaminase
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001012732.1 |
799 |
Missense Mutation |
CAG,GAG |
Q189E |
NP_001012750.1 |
NM_001921.2 |
799 |
Missense Mutation |
CAG,GAG |
Q178E |
NP_001912.2 |
XM_005262778.3 |
799 |
Missense Mutation |
CAG,GAG |
Q189E |
XP_005262835.1 |
XM_005262779.3 |
799 |
Missense Mutation |
CAG,GAG |
Q178E |
XP_005262836.1 |
XM_005262780.3 |
799 |
Missense Mutation |
CAG,GAG |
Q178E |
XP_005262837.1 |
XM_005262781.4 |
799 |
Missense Mutation |
CAG,GAG |
Q119E |
XP_005262838.1 |
XM_005262782.3 |
799 |
Missense Mutation |
CAG,GAG |
Q119E |
XP_005262839.1 |
XM_006714115.3 |
799 |
Missense Mutation |
CAG,GAG |
Q119E |
XP_006714178.1 |
XM_006714116.3 |
799 |
Missense Mutation |
CAG,GAG |
Q119E |
XP_006714179.1 |
XM_011531674.2 |
799 |
Missense Mutation |
CAG,GAG |
Q178E |
XP_011529976.1 |
XM_011531675.2 |
799 |
Missense Mutation |
CAG,GAG |
Q119E |
XP_011529977.1 |
XM_011531676.2 |
799 |
Missense Mutation |
CAG,GAG |
Q119E |
XP_011529978.1 |
XM_017007816.1 |
799 |
Missense Mutation |
CAG,GAG |
Q178E |
XP_016863305.1 |
XM_017007817.1 |
799 |
Missense Mutation |
CAG,GAG |
Q178E |
XP_016863306.1 |
XM_017007818.1 |
799 |
Missense Mutation |
CAG,GAG |
Q178E |
XP_016863307.1 |
XM_017007819.1 |
799 |
Missense Mutation |
CAG,GAG |
Q178E |
XP_016863308.1 |
XM_017007820.1 |
799 |
Missense Mutation |
CAG,GAG |
Q178E |
XP_016863309.1 |
XM_017007821.1 |
799 |
Missense Mutation |
CAG,GAG |
Q178E |
XP_016863310.1 |
XM_017007822.1 |
799 |
Missense Mutation |
CAG,GAG |
Q178E |
XP_016863311.1 |
XM_017007823.1 |
799 |
Missense Mutation |
CAG,GAG |
Q119E |
XP_016863312.1 |
XM_017007824.1 |
799 |
Missense Mutation |
CAG,GAG |
Q119E |
XP_016863313.1 |
View Full Product Details