Product Details

SNP ID
rs143249561
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:152769473 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGATGTACAATACATAGTTACTTA[A/G]GTGATAAAGAGTTAATAAAATTTTC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TIGD4 PubMed Links

Gene Details

Gene
TIGD4
Gene Name
tigger transposable element derived 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145720.3 2353 Missense Mutation CCT,CTT P511L NP_663772.1
XM_005262807.4 2353 Missense Mutation CCT,CTT P511L XP_005262864.1

View Full Product Details