Product Details

SNP ID
rs145023591
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:72283295 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGCTGAACTGAGGTGGACCCTCTT[G/T]GTGGGTGGGGAGGATGGTACGGTGA
Phenotype
MIM: 605011
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ADAMTS3 PubMed Links

Gene Details

Gene
ADAMTS3
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014243.2 4027 Silent Mutation NP_055058.2
XM_011532421.2 4027 Silent Mutation XP_011530723.1
XM_011532422.2 4027 Silent Mutation XP_011530724.1

View Full Product Details