Product Details

SNP ID
rs145522455
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:155672785 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGGCAATGGTTGGACTCTGTCAA[C/T]ACTTCTCTTCATCACAATATTTTTA
Phenotype
MIM: 139396
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GUCY1A3 PubMed Links

Gene Details

Gene
GUCY1A3
Gene Name
guanylate cyclase 1 soluble subunit alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000856.5 Intron NP_000847.2
NM_001130682.2 Intron NP_001124154.1
NM_001130683.3 Intron NP_001124155.1
NM_001130684.2 Intron NP_001124156.1
NM_001130685.2 Intron NP_001124157.1
NM_001130687.2 Intron NP_001124159.1
NM_001256449.1 Intron NP_001243378.1
XM_005262955.2 Intron XP_005263012.1
XM_005262956.2 Intron XP_005263013.1
XM_005262957.2 Intron XP_005263014.1
XM_006714196.2 Intron XP_006714259.1
XM_006714197.2 Intron XP_006714260.1
XM_006714198.2 Intron XP_006714261.1
XM_011531900.2 Intron XP_011530202.1

View Full Product Details