Product Details

SNP ID
rs145746519
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:56911046 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGTGCTCCAGATATTTACAGTTC[A/T]AATAAAGATCTTCCCCCTGAAACAC
Phenotype
MIM: 600571
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
REST PubMed Links

Gene Details

Gene
REST
Gene Name
RE1 silencing transcription factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193508.1 716 Silent Mutation TCA,TCT S136S NP_001180437.1
NM_005612.4 716 Silent Mutation TCA,TCT S136S NP_005603.3
XM_011534401.2 716 Silent Mutation TCA,TCT S136S XP_011532703.1
XM_017008527.1 716 Silent Mutation TCA,TCT S136S XP_016864016.1

View Full Product Details