Product Details

SNP ID
rs146860744
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.4:25123708 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATATTCTAACAATTAGAAAAATG[C/T]TAATTGTATATTATAACCTGTTAAG
Phenotype
MIM: 613009
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
SEPSECS PubMed Links

Gene Details

Gene
SEPSECS
Gene Name
Sep (O-phosphoserine) tRNA:Sec (selenocysteine) tRNA synthase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016955.3 1916 UTR 3 NP_058651.3
XM_011513846.2 1916 UTR 3 XP_011512148.1
XM_011513847.2 1916 UTR 3 XP_011512149.1
XM_011513848.1 1916 UTR 3 XP_011512150.1
XM_017008277.1 1916 UTR 3 XP_016863766.1
XM_017008278.1 1916 UTR 3 XP_016863767.1

View Full Product Details