Product Details

SNP ID
rs147428278
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:140388973 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATCTCCAAACAACTAAAACTGTGT[A/G]ACTTCTAATGGTATTAATCTCTTAG
Phenotype
MIM: 601858
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CLGN PubMed Links

Gene Details

Gene
CLGN
Gene Name
calmegin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130675.1 2218 UTR 3 NP_001124147.1
NM_004362.2 2218 UTR 3 NP_004353.1
Gene
SCOC
Gene Name
short coiled-coil protein
There are no transcripts associated with this gene.

View Full Product Details