Product Details

SNP ID
rs147447027
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:25121306 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACATTGTGTAGTGACCAAAAGAAT[A/G]TTCTATGTTCCACCATTTTCTTTTC
Phenotype
MIM: 613009
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SEPSECS PubMed Links

Gene Details

Gene
SEPSECS
Gene Name
Sep (O-phosphoserine) tRNA:Sec (selenocysteine) tRNA synthase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016955.3 4318 UTR 3 NP_058651.3
XM_011513846.2 4318 UTR 3 XP_011512148.1
XM_011513847.2 4318 UTR 3 XP_011512149.1
XM_011513848.1 4318 UTR 3 XP_011512150.1
XM_017008277.1 4318 UTR 3 XP_016863766.1
XM_017008278.1 4318 UTR 3 XP_016863767.1

View Full Product Details