Product Details

SNP ID
rs147614165
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:105683151 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCCACAGGTTTCTGGTTAGCTGAC[A/G]AAGTAGCAGGTTTCCCAGTATTGTT
Phenotype
MIM: 611355
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARHGEF38 PubMed Links

Gene Details

Gene
ARHGEF38
Gene Name
Rho guanine nucleotide exchange factor 38
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242729.1 1878 Intron NP_001229658.1
NM_017700.1 1878 Intron NP_060170.1
XM_006714244.3 1878 Intron XP_006714307.1
XM_011532050.2 1878 Intron XP_011530352.1
XM_011532051.2 1878 Intron XP_011530353.1
Gene
INTS12
Gene Name
integrator complex subunit 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142471.1 1878 Missense Mutation TCG,TTG S324L NP_001135943.1
NM_020395.3 1878 Missense Mutation TCG,TTG S324L NP_065128.2
XM_005263148.4 1878 Missense Mutation TCG,TTG S324L XP_005263205.1
XM_011532143.2 1878 Missense Mutation TCG,TTG S324L XP_011530445.1
XM_011532145.2 1878 Missense Mutation TCG,TTG S324L XP_011530447.1

View Full Product Details