Product Details

SNP ID
rs148305209
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:25662723 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTTCAGCAGATAACACTGAGGCAC[C/G]TGTAACCAAGATTGAACTTCTGCCG
Phenotype
MIM: 604217
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SLC34A2 PubMed Links

Gene Details

Gene
SLC34A2
Gene Name
solute carrier family 34 member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001177998.1 232 Missense Mutation CCT,CGT P43R NP_001171469.1
NM_001177999.1 232 Missense Mutation CCT,CGT P43R NP_001171470.1
NM_006424.2 232 Missense Mutation CCT,CGT P44R NP_006415.2

View Full Product Details