Product Details

SNP ID
rs148888761
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:25923841 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACCACATAATCGAATGAAATGCG[C/T]AGGCAGACTGGGTCTTGTTTTCTAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SMIM20 PubMed Links

Gene Details

Gene
SMIM20
Gene Name
small integral membrane protein 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145432.1 Intron NP_001138904.1

View Full Product Details