Product Details

SNP ID
rs150105001
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:158680554 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTCTTCAACTTCTACTGTGCCTC[A/G]AATTACTACCCATTATACTATTTAT
Phenotype
MIM: 616210 MIM: 231675
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C4orf46 PubMed Links

Gene Details

Gene
C4orf46
Gene Name
chromosome 4 open reading frame 46
There are no transcripts associated with this gene.

Gene
ETFDH
Gene Name
electron transfer flavoprotein dehydrogenase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001281737.1 478 Intron NP_001268666.1
NM_001281738.1 478 Intron NP_001268667.1
NM_004453.3 478 Missense Mutation CAA,CGA Q41R NP_004444.2

View Full Product Details