Product Details

SNP ID
rs150319742
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:82631385 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCTCGATCATCGGCTTGGTTGCC[C/T]GTTTGCGGTCAGTGGCCAGCCCCAG
Phenotype
MIM: 608370
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC00575 PubMed Links

Gene Details

Gene
LINC00575
Gene Name
long intergenic non-protein coding RNA 575
There are no transcripts associated with this gene.

Gene
SCD5
Gene Name
stearoyl-CoA desaturase 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037582.2 1255 Missense Mutation NP_001032671.2
NM_024906.2 1255 Intron NP_079182.2

View Full Product Details