Product Details

SNP ID
rs150510400
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:56911355 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATACAGCACACCTGAAACACCACAC[C/T]AGAGCTGGGGATAATGAGCGAGTCT
Phenotype
MIM: 600571
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
REST PubMed Links

Gene Details

Gene
REST
Gene Name
RE1 silencing transcription factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193508.1 1025 Silent Mutation ACC,ACT T239T NP_001180437.1
NM_005612.4 1025 Silent Mutation ACC,ACT T239T NP_005603.3
XM_011534401.2 1025 Silent Mutation ACC,ACT T239T XP_011532703.1
XM_017008527.1 1025 Silent Mutation ACC,ACT T239T XP_016864016.1

View Full Product Details