Product Details

SNP ID
rs150967429
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:174643970 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTGGCATTACCTGGACAGGGTGG[C/T]TGGGGCCCTTTGGAGTCATGCCATC
Phenotype
MIM: 600421
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GLRA3 PubMed Links

Gene Details

Gene
GLRA3
Gene Name
glycine receptor alpha 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042543.2 1715 Missense Mutation AAC,AGC N389S NP_001036008.1
NM_006529.3 1715 Missense Mutation AAC,AGC N404S NP_006520.2
XM_011532267.2 1715 Intron XP_011530569.1
XM_011532268.2 1715 Missense Mutation AAC,AGC N209S XP_011530570.1
XM_017008630.1 1715 Missense Mutation AAC,AGC N364S XP_016864119.1

View Full Product Details