Product Details
- SNP ID
-
rs138407792
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:148826906 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCCATGCGCCGGACCACGACGTCAC[A/G]CAGGAAAGGGACGAGGTGTGGGTGG
- Phenotype
-
MIM: 109690
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ADRB2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs1042714] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ADRB2
- Gene Name
- adrenoceptor beta 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000024.5 |
314 |
Silent Mutation |
ACA,ACG |
T25T |
NP_000015.1 |
View Full Product Details