Product Details

SNP ID
rs138944821
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:1201678 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGAGGCTCGTGCTGCCCAACCCC[A/G]GCCTAGACGCCCGGATCCCGTCCCT
Phenotype
MIM: 608893
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC6A19 PubMed Links

Gene Details

Gene
SLC6A19
Gene Name
solute carrier family 6 member 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001003841.2 84 Missense Mutation AGC,GGC S10G NP_001003841.1

View Full Product Details