Product Details

SNP ID
rs139043819
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:146514988 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAAGGATGGGAATCTCTGCCTTCT[C/T]AGTTTTCCCTTTGCCAGAGGAGGGA
Phenotype
MIM: 605409
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GPR151 PubMed Links

Gene Details

Gene
GPR151
Gene Name
G protein-coupled receptor 151
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_194251.2 1126 Missense Mutation AAG,GAG K376E NP_919227.2
Gene
TCERG1
Gene Name
transcription elongation regulator 1
There are no transcripts associated with this gene.

View Full Product Details