Product Details

SNP ID
rs139263174
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:73173660 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCATGGGGCCACTGTGGTGCAGTA[C/T]ATCCCTCCTCCTTATGGTTCTCCAG
Phenotype
MIM: 614909
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TMEM174 PubMed Links

Gene Details

Gene
TMEM174
Gene Name
transmembrane protein 174
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153217.2 465 Silent Mutation TAC,TAT Y139Y NP_694949.1

View Full Product Details