Product Details
- SNP ID
-
rs139624975
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:81428635 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAATTTCCCCCCATTTCGCCATCAT[C/G]CCTTGGAGTGCCCGGTTGATTACTC
- Phenotype
-
MIM: 607389
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
SSBP2
PubMed Links
Gene Details
- Gene
- SSBP2
- Gene Name
- single stranded DNA binding protein 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001256732.1 |
1532 |
Missense Mutation |
CAT,GAT |
H344D |
NP_001243661.1 |
NM_001256733.1 |
1532 |
Missense Mutation |
CAT,GAT |
H316D |
NP_001243662.1 |
NM_001256734.1 |
1532 |
Missense Mutation |
CAT,GAT |
H314D |
NP_001243663.1 |
NM_001256735.1 |
1532 |
Missense Mutation |
CAT,GAT |
H306D |
NP_001243664.1 |
NM_001256736.1 |
1532 |
Intron |
|
|
NP_001243665.1 |
NM_012446.3 |
1532 |
Missense Mutation |
CAT,GAT |
H336D |
NP_036578.2 |
XM_011543297.2 |
1532 |
Intron |
|
|
XP_011541599.1 |
XM_017009304.1 |
1532 |
Missense Mutation |
CAT,GAT |
H340D |
XP_016864793.1 |
XM_017009305.1 |
1532 |
Missense Mutation |
CAT,GAT |
H332D |
XP_016864794.1 |
XM_017009306.1 |
1532 |
Intron |
|
|
XP_016864795.1 |
XM_017009307.1 |
1532 |
Missense Mutation |
CAT,GAT |
H302D |
XP_016864796.1 |
XM_017009308.1 |
1532 |
Missense Mutation |
CAT,GAT |
H232D |
XP_016864797.1 |
XM_017009309.1 |
1532 |
Missense Mutation |
CAT,GAT |
H232D |
XP_016864798.1 |
XM_017009310.1 |
1532 |
Missense Mutation |
CAT,GAT |
H232D |
XP_016864799.1 |
View Full Product Details