Product Details
- SNP ID
-
rs140990806
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:140363757 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTGGGGGCCTTATCCAGGACGTGC[A/G]CAGGAAGGTCCCGTGGTACCCCAGC
- Phenotype
-
MIM: 610207
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SLC4A9
PubMed Links
Gene Details
- Gene
- SLC4A9
- Gene Name
- solute carrier family 4 member 9
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001258426.1 |
1144 |
Intron |
|
|
NP_001245355.1 |
NM_001258427.1 |
1144 |
Missense Mutation |
CAC,CGC |
H370R |
NP_001245356.1 |
NM_001258428.1 |
1144 |
Missense Mutation |
CAC,CGC |
H394R |
NP_001245357.1 |
NM_031467.2 |
1144 |
Missense Mutation |
CAC,CGC |
H370R |
NP_113655.2 |
XM_005268521.3 |
1144 |
Missense Mutation |
CAC,CGC |
H394R |
XP_005268578.2 |
XM_011537694.2 |
1144 |
Missense Mutation |
CAC,CGC |
H418R |
XP_011535996.1 |
XM_017009933.1 |
1144 |
Missense Mutation |
CAC,CGC |
H418R |
XP_016865422.1 |
XM_017009934.1 |
1144 |
Missense Mutation |
CAC,CGC |
H418R |
XP_016865423.1 |
XM_017009935.1 |
1144 |
Intron |
|
|
XP_016865424.1 |
XM_017009936.1 |
1144 |
Missense Mutation |
CAC,CGC |
H394R |
XP_016865425.1 |
XM_017009937.1 |
1144 |
Missense Mutation |
CAC,CGC |
H380R |
XP_016865426.1 |
XM_017009938.1 |
1144 |
Missense Mutation |
CAC,CGC |
H418R |
XP_016865427.1 |
XM_017009939.1 |
1144 |
Intron |
|
|
XP_016865428.1 |
XM_017009940.1 |
1144 |
Missense Mutation |
CAC,CGC |
H418R |
XP_016865429.1 |
XM_017009941.1 |
1144 |
Missense Mutation |
CAC,CGC |
H418R |
XP_016865430.1 |
XM_017009942.1 |
1144 |
Intron |
|
|
XP_016865431.1 |
XM_017009943.1 |
1144 |
Intron |
|
|
XP_016865432.1 |
View Full Product Details