Product Details

SNP ID
rs141212131
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:3599686 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGACCAGAGCAACGAGGATGACGA[A/G]GACAAGGCCGAGGCTCCGCACGCGC
Phenotype
MIM: 606197
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IRX1 PubMed Links

Gene Details

Gene
IRX1
Gene Name
iroquois homeobox 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024337.3 790 Silent Mutation GAA,GAG E246E NP_077313.3
XM_017009845.1 790 Silent Mutation GAA,GAG E246E XP_016865334.1

View Full Product Details