Product Details
- SNP ID
-
rs142007771
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:121852097 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCCTTGGACCCCAGGCAGATCGCCC[A/C]CCGCCGCCCCCTGGCCGCAGCCGCC
- Phenotype
-
MIM: 608847
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
FTMT
PubMed Links
Gene Details
- Gene
- FTMT
- Gene Name
- ferritin mitochondrial
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_177478.1 |
143 |
Missense Mutation |
CAC,CCC |
H45P |
NP_803431.1 |
View Full Product Details