Product Details
- SNP ID
-
rs142545546
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:176908564 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTCTGCTGCCACTTCCCCTCCACAG[C/T]TGAACATGCTCTTCCACTCCCTTCA
- Phenotype
-
MIM: 142570
MIM: 609433
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
HK3
PubMed Links
Gene Details
- Gene
- HK3
- Gene Name
- hexokinase 3
There are no transcripts associated with this gene.
- Gene
- UIMC1
- Gene Name
- ubiquitin interaction motif containing 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001199297.2 |
1084 |
Missense Mutation |
ACT,GCT |
T603A |
NP_001186226.1 |
NM_001199298.1 |
1084 |
Missense Mutation |
ACT,GCT |
T603A |
NP_001186227.1 |
NM_001317961.1 |
1084 |
Missense Mutation |
ACT,GCT |
T437A |
NP_001304890.1 |
NM_016290.4 |
1084 |
Missense Mutation |
ACT,GCT |
T603A |
NP_057374.3 |
XM_005265930.2 |
1084 |
Missense Mutation |
ACT,GCT |
T603A |
XP_005265987.1 |
XM_005265932.2 |
1084 |
Missense Mutation |
ACT,GCT |
T322A |
XP_005265989.1 |
XM_005265933.2 |
1084 |
Missense Mutation |
ACT,GCT |
T322A |
XP_005265990.1 |
XM_005265934.2 |
1084 |
Missense Mutation |
ACT,GCT |
T300A |
XP_005265991.1 |
XM_005265935.2 |
1084 |
Missense Mutation |
ACT,GCT |
T300A |
XP_005265992.1 |
XM_005265936.2 |
1084 |
Missense Mutation |
ACT,GCT |
T233A |
XP_005265993.1 |
XM_006714871.2 |
1084 |
Missense Mutation |
ACT,GCT |
T603A |
XP_006714934.1 |
XM_006714874.2 |
1084 |
Missense Mutation |
ACT,GCT |
T300A |
XP_006714937.1 |
XM_011534568.2 |
1084 |
Intron |
|
|
XP_011532870.1 |
XM_011534570.1 |
1084 |
Intron |
|
|
XP_011532872.1 |
XM_017009574.1 |
1084 |
Missense Mutation |
ACT,GCT |
T525A |
XP_016865063.1 |
XM_017009575.1 |
1084 |
Missense Mutation |
ACT,GCT |
T504A |
XP_016865064.1 |
XM_017009576.1 |
1084 |
Missense Mutation |
ACT,GCT |
T322A |
XP_016865065.1 |
XM_017009577.1 |
1084 |
Missense Mutation |
ACT,GCT |
T322A |
XP_016865066.1 |
XM_017009578.1 |
1084 |
Missense Mutation |
ACT,GCT |
T233A |
XP_016865067.1 |
XM_017009579.1 |
1084 |
Missense Mutation |
ACT,GCT |
T233A |
XP_016865068.1 |
View Full Product Details