Product Details

SNP ID
rs142629562
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:94157229 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATTTTGGTAGCCTGCCCTTTTGTA[G/T]TATTGCTGAATTCCCCCACCTGCCT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
KIAA0825 PubMed Links

Gene Details

Gene
KIAA0825
Gene Name
KIAA0825
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145678.1 Intron NP_001139150.1
NM_173665.2 Intron NP_775936.1
XM_006714588.2 Intron XP_006714651.1
XM_011543324.2 Intron XP_011541626.1
XM_011543325.2 Intron XP_011541627.1
XM_011543327.2 Intron XP_011541629.1
XM_011543328.2 Intron XP_011541630.1
XM_011543329.2 Intron XP_011541631.1
XM_011543330.2 Intron XP_011541632.1
XM_011543331.1 Intron XP_011541633.1
XM_011543332.1 Intron XP_011541634.1
XM_011543333.2 Intron XP_011541635.1
XM_011543334.2 Intron XP_011541636.1
XM_011543335.2 Intron XP_011541637.1
XM_017009373.1 Intron XP_016864862.1
XM_017009374.1 Intron XP_016864863.1
XM_017009375.1 Intron XP_016864864.1
XM_017009376.1 Intron XP_016864865.1
XM_017009377.1 Intron XP_016864866.1

View Full Product Details