Product Details
- SNP ID
-
rs142841909
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:170108145 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGGAAGAGAAAATCAGATGTTTCCT[C/T]TAGCACAGCCTCCTTGGCCTTAGAG
- Phenotype
-
MIM: 601093
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
FOXI1
PubMed Links
Gene Details
- Gene
- FOXI1
- Gene Name
- forkhead box I1
View Full Product Details