Product Details

SNP ID
rs143047953
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:65669011 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATTTCATTATTAATAAACAAAAAA[C/T]ATACTCTAGTTGGAATTAAACTCTC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SGTB PubMed Links

Gene Details

Gene
SGTB
Gene Name
small glutamine rich tetratricopeptide repeat containing beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019072.2 2506 UTR 3 NP_061945.1
XM_005248548.3 2506 UTR 3 XP_005248605.1
XM_017009597.1 2506 UTR 3 XP_016865086.1
Gene
TRAPPC13
Gene Name
trafficking protein particle complex 13
There are no transcripts associated with this gene.

View Full Product Details