Product Details

SNP ID
rs143233559
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:132696677 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATTACAGTTTCAGGCTTTGCAGAA[C/T]GCTTTCTGTTTGGAGAAGAAAAAAA
Phenotype
MIM: 604683
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KIF3A PubMed Links

Gene Details

Gene
KIF3A
Gene Name
kinesin family member 3A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300791.1 2297 Missense Mutation CAT,CGT H713R NP_001287720.1
NM_001300792.1 2297 Missense Mutation CAT,CGT H689R NP_001287721.1
NM_007054.6 2297 Missense Mutation CAT,CGT H686R NP_008985.3
XM_006714526.3 2297 Intron XP_006714589.1
XM_017008995.1 2297 Intron XP_016864484.1
XM_017008996.1 2297 Intron XP_016864485.1

View Full Product Details