Product Details

SNP ID
rs143716622
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:160393840 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGACTTTGTTTTGAAATGTAAAA[A/G]TGATGAAAATCCCAACTCACAAAGG
Phenotype
MIM: 605974 MIM: 615253
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLU7 PubMed Links

Gene Details

Gene
SLU7
Gene Name
SLU7 homolog, splicing factor
There are no transcripts associated with this gene.

Gene
ZBED8
Gene Name
zinc finger BED-type containing 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303251.1 2233 Missense Mutation CTT,TTT L551F NP_001290180.1
NM_022090.4 2233 Missense Mutation CTT,TTT L551F NP_071373.2

View Full Product Details