Product Details
- SNP ID
-
rs143922310
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:26881276 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAACTGAGCGAATCTGCTATGGAAT[C/T]ATTCCCTTCATAGGCATACGTTGCC
- Phenotype
-
MIM: 609974
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CDH9
PubMed Links
Gene Details
- Gene
- CDH9
- Gene Name
- cadherin 9
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_016279.3 |
2399 |
Missense Mutation |
AAT,GAT |
N744D |
NP_057363.3 |
View Full Product Details