Product Details

SNP ID
rs144032011
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:140666585 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGCCATGGTACCTTTCCCTTAAT[A/G]ATGAATGGTTGGTAACCATATGTCC
Phenotype
MIM: 609385 MIM: 142810 MIM: 600549
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DND1 PubMed Links

Gene Details

Gene
DND1
Gene Name
DND microRNA-mediated repression inhibitor 1
There are no transcripts associated with this gene.

Gene
HARS
Gene Name
histidyl-tRNA synthetase
There are no transcripts associated with this gene.

Gene
IK
Gene Name
IK cytokine, down-regulator of HLA II
There are no transcripts associated with this gene.

Gene
WDR55
Gene Name
WD repeat domain 55
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017706.4 Intron NP_060176.2
XM_005268469.2 Intron XP_005268526.1
XM_017009600.1 Intron XP_016865089.1

View Full Product Details