Product Details

SNP ID
rs144283422
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:41739420 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATCACACCCAGTACTCTTCTGTAC[A/G]TCATCCACTGTCAGGCCTTCCCAGA
Phenotype
MIM: 601424
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OXCT1 PubMed Links

Gene Details

Gene
OXCT1
Gene Name
3-oxoacid CoA-transferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000436.3 1822 Silent Mutation NP_000427.1
XM_017009494.1 1822 Silent Mutation XP_016864983.1

View Full Product Details