Product Details

SNP ID
rs144332148
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:115524477 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCATCCAGCAAGTCACTAGCAGTT[C/T]GTTTGTGCAAGTTTGTGGCATCAAA
Phenotype
MIM: 608767
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FEM1C PubMed Links

Gene Details

Gene
FEM1C
Gene Name
fem-1 homolog C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020177.2 2333 Missense Mutation CAA,CGA Q562R NP_064562.1
XM_005272035.4 2333 Missense Mutation CAA,CGA Q562R XP_005272092.1
XM_017009647.1 2333 Missense Mutation CAA,CGA Q409R XP_016865136.1

View Full Product Details