Product Details

SNP ID
rs144757190
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:131651879 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGAAACAAGACTGGAAACCAATA[C/T]TTCCTTTCCCAATTTATTATCTGTC
Phenotype
MIM: 610594
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FNIP1 PubMed Links

Gene Details

Gene
FNIP1
Gene Name
folliculin interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008738.2 3287 Missense Mutation ATA,GTA I1049V NP_001008738.2
NM_133372.2 3287 Missense Mutation ATA,GTA I1077V NP_588613.2

View Full Product Details