Product Details

SNP ID
rs145345185
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:71011335 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCTCCTTCTTCCCAATTTCCTAAA[C/T]ATCCACCACAGGAAAAACACTGTAC
Phenotype
MIM: 600355
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NAIP PubMed Links

Gene Details

Gene
NAIP
Gene Name
NLR family apoptosis inhibitory protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004536.2 1326 Missense Mutation TAT,TGT Y203C NP_004527.2
NM_022892.1 1326 Intron NP_075043.1
XM_011543413.2 1326 Intron XP_011541715.1
XM_017009490.1 1326 Intron XP_016864979.1

View Full Product Details