Product Details

SNP ID
rs145512953
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:156920478 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATAGATTACCTTGTGTGTTTCTG[C/T]GAACAATAGGTTTCCATGAGTTTCC
Phenotype
MIM: 610096
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TIMD4 PubMed Links

Gene Details

Gene
TIMD4
Gene Name
T-cell immunoglobulin and mucin domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146726.1 1011 Silent Mutation TCA,TCG S318S NP_001140198.1
NM_138379.2 1011 Silent Mutation TCA,TCG S346S NP_612388.2
XM_011534694.2 1011 Intron XP_011532996.1
XM_017010021.1 1011 Silent Mutation TCA,TCG S291S XP_016865510.1

View Full Product Details