Product Details

SNP ID
rs146317412
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:139398025 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCACATAGTAGGTATACAAAAAGT[A/G]TATTTGCAGACCGAATGAACGGGAT
Phenotype
MIM: 609447
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MZB1 PubMed Links

Gene Details

Gene
MZB1
Gene Name
marginal zone B and B1 cell specific protein
There are no transcripts associated with this gene.

Gene
PROB1
Gene Name
proline rich basic protein 1
There are no transcripts associated with this gene.

Gene
SPATA24
Gene Name
spermatogenesis associated 24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_194296.1 Intron NP_919272.1
XM_005271916.4 Intron XP_005271973.1
XM_011543252.2 Intron XP_011541554.1
XM_011543253.2 Intron XP_011541555.1
XM_011543254.2 Intron XP_011541556.1

View Full Product Details