Product Details

SNP ID
rs147233991
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.5:81420330 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACTCCGTACAGTTGTTTGAATCA[C/T]ATTTGGACCCGCTTTCTTCACAAAA
Phenotype
MIM: 607389
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
SSBP2 PubMed Links

Gene Details

Gene
SSBP2
Gene Name
single stranded DNA binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256732.1 1282 UTR 3 NP_001243661.1
NM_001256733.1 1282 UTR 3 NP_001243662.1
NM_001256734.1 1282 UTR 3 NP_001243663.1
NM_001256735.1 1282 UTR 3 NP_001243664.1
NM_001256736.1 1282 UTR 3 NP_001243665.1
NM_012446.3 1282 UTR 3 NP_036578.2
XM_011543297.2 1282 UTR 3 XP_011541599.1
XM_017009304.1 1282 UTR 3 XP_016864793.1
XM_017009305.1 1282 UTR 3 XP_016864794.1
XM_017009306.1 1282 UTR 3 XP_016864795.1
XM_017009307.1 1282 UTR 3 XP_016864796.1
XM_017009308.1 1282 UTR 3 XP_016864797.1
XM_017009309.1 1282 UTR 3 XP_016864798.1
XM_017009310.1 1282 UTR 3 XP_016864799.1

View Full Product Details