Product Details

SNP ID
rs147770445
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:176371791 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGTCTACAGGCTATCGATAACCAG[C/T]GGGAGGTTTTCCTCTTGGCAGCCAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARL10 PubMed Links

Gene Details

Gene
ARL10
Gene Name
ADP ribosylation factor like GTPase 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317948.1 727 Intron NP_001304877.1
NM_173664.5 727 Silent Mutation CGG,TGG R211W NP_775935.1
XM_011534529.2 727 Intron XP_011532831.1
XM_011534530.2 727 Intron XP_011532832.1
XM_011534531.2 727 Intron XP_011532833.1
XM_017009372.1 727 Intron XP_016864861.1
Gene
KIAA1191
Gene Name
KIAA1191
There are no transcripts associated with this gene.

Gene
MIR1271
Gene Name
microRNA 1271
There are no transcripts associated with this gene.

View Full Product Details