Product Details
- SNP ID
-
rs148718250
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:141135809 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTCAAACTGATCTGCAGCTCACAGA[C/T]ATAAATGACCATGCCCCAGAGTTCC
- Phenotype
-
MIM: 606331
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PCDHB5
PubMed Links
Gene Details
- Gene
- PCDHB5
- Gene Name
- protocadherin beta 5
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_015669.4 |
604 |
Silent Mutation |
GAC,GAT |
D125D |
NP_056484.2 |
View Full Product Details