Product Details

SNP ID
rs148757696
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:137802407 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGACCAAATAAACTAGAAATATTG[C/T]CTTGTATACGATTTAGGAAGTAGAC
Phenotype
MIM: 601770
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NPY6R PubMed Links

Gene Details

Gene
NPY6R
Gene Name
neuropeptide Y receptor Y6 (pseudogene)
There are no transcripts associated with this gene.

View Full Product Details