Product Details

SNP ID
rs149605593
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:179028048 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGAACAGGGATTCTCTTTTCCAA[A/G]CCAAAGGTCATCTCCCAGTTAGAGC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF879 PubMed Links

Gene Details

Gene
ZNF879
Gene Name
zinc finger protein 879
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136116.1 487 Silent Mutation AAA,AAG K59K NP_001129588.1
XM_005265908.3 487 Silent Mutation AAA,AAG K59K XP_005265965.1
XM_011534550.2 487 Silent Mutation AAA,AAG K59K XP_011532852.1
XM_011534551.2 487 Silent Mutation AAA,AAG K59K XP_011532853.1

View Full Product Details