Product Details

SNP ID
rs149628976
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:90566258 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGCCTGATACAAGGTCACAAAAAT[G/T]TATACCCATGCTTTCATCTAAGAGT
Phenotype
MIM: 602851
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ADGRV1 PubMed Links

Gene Details

Gene
ADGRV1
Gene Name
adhesion G protein-coupled receptor V1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032119.3 Intron NP_115495.3
XM_017009963.1 Intron XP_016865452.1
XM_017009964.1 Intron XP_016865453.1
XM_017009965.1 Intron XP_016865454.1
XM_017009966.1 Intron XP_016865455.1
XM_017009967.1 Intron XP_016865456.1
XM_017009968.1 Intron XP_016865457.1
XM_017009969.1 Intron XP_016865458.1
XM_017009970.1 Intron XP_016865459.1
XM_017009971.1 Intron XP_016865460.1
XM_017009972.1 Intron XP_016865461.1
XM_017009973.1 Intron XP_016865462.1
XM_017009974.1 Intron XP_016865463.1

View Full Product Details