Product Details

SNP ID
rs149889700
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:160010783 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAAGGTTGAGAACAAATCTAATGA[A/G]GATGTGAATTCCTCTGAACTAGATG
Phenotype
MIM: 601963
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TTC1 PubMed Links

Gene Details

Gene
TTC1
Gene Name
tetratricopeptide repeat domain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282500.1 375 Silent Mutation GAA,GAG E85E NP_001269429.1
NM_003314.2 375 Silent Mutation GAA,GAG E85E NP_003305.1

View Full Product Details